Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
casp7
Entrez Gene ID
553634
Gene Name
caspase 7, apoptosis-related cysteine peptidase
Gene Aliases
zgc:110595
Chromosome Location
Chr.12: 30569619 - 30583668 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020607.1 NP_001018443.1 3 - 4 383 100
GenBank mRNA BC095327.1 - 3 - 4 383 100
MG957999.1 - 2 - 3 242 100

View Full Product Details