Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
ora2
Entrez Gene ID
100005331
Gene Name
olfactory receptor class A related 2
Gene Aliases
v1r1
Chromosome Location
Chr.22: 213650 - 214561 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098395.1 NP_001091865.1 1 - 1 857 70
GenBank mRNA AM778164.1 - 1 - 1 318 70

View Full Product Details