Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
setd7
Entrez Gene ID
436729
Gene Name
SET domain containing 7, histone lysine methyltransferase
Gene Aliases
set7, zgc:101860, zgc:92330
Chromosome Location
Chr.14: 46199941 - 46213584 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002456.1 NP_001002456.1 4 - 5 643 84
XM_005169609.3 XP_005169666.1 3 - 4 556 84
GenBank mRNA BC076014.1 - 4 - 5 643 84

View Full Product Details