Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cd99
Entrez Gene ID
559896
Gene Name
CD99 molecule
Gene Aliases
im:7159661, zgc:153928
Chromosome Location
Chr.1: 33355377 - 33380072 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079980.1 NP_001073449.1 6 - 7 278 153
XM_005159923.4 XP_005159980.1 6 - 7 362 153
GenBank mRNA BC125914.1 - 6 - 7 278 153

View Full Product Details