Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gosr2
Entrez Gene ID
324569
Gene Name
golgi SNAP receptor complex member 2
Gene Aliases
wu:fc33b02, zgc:64068
Chromosome Location
Chr.3: 37649557 - 37660860 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199688.1 NP_955982.1 4 - 5 377 122
GenBank mRNA BC053235.1 - 4 - 5 377 122
BC165142.1 - 4 - 5 375 122

View Full Product Details