Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nr1d1
Entrez Gene ID
494487
Gene Name
nuclear receptor subfamily 1, group d, member 1
Gene Aliases
Eip75B, zrev-erba
Chromosome Location
Chr.3: 23488621 - 23496223 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205729.2 NP_991292.2 7 - 8 2009 96
GenBank mRNA AY391444.1 - 7 - 8 1810 96

View Full Product Details