Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gnb3b
Entrez Gene ID
406483
Gene Name
guanine nucleotide binding protein (G protein), beta polypeptide 3b
Gene Aliases
gnb3, wu:fk54b04, zgc:73058, zgc:77780
Chromosome Location
Chr.19: 5106226 - 5125626 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213202.1 NP_998367.1 6 - 7 676 77
XM_005159345.4 XP_005159402.1 6 - 7 685 77
GenBank mRNA BC059436.1 - 6 - 7 657 77
BC065866.1 - 6 - 7 676 77

View Full Product Details