Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
psmd8
Entrez Gene ID
415221
Gene Name
proteasome 26S subunit, non-ATPase 8
Gene Aliases
fa93g07, fb17c09, fb49a10, wu:fa93g07, wu:fb17c09, wu:fb49a10, zgc:86762
Chromosome Location
Chr.18: 44615596 - 44623646 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002131.1 NP_001002131.1 5 - 6 614 109
GenBank mRNA BC071432.1 - 5 - 6 614 109

View Full Product Details