Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
gli2b
Entrez Gene ID
548610
Gene Name
GLI family zinc finger 2b
Gene Aliases
-
Chromosome Location
Chr.11: 43550194 - 43713750 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015069.1 NP_001015069.1 2 - 3 441 77
GenBank mRNA AY928397.2 - 2 - 3 441 77
BC162463.1 - 2 - 3 238 77

View Full Product Details