Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dpf2
Entrez Gene ID
474324
Gene Name
D4, zinc and double PHD fingers family 2
Gene Aliases
im:7146797, zgc:101789
Chromosome Location
Chr.5: 37744600 - 37759706 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007152.1 NP_001007153.1 6 - 7 663 66
GenBank mRNA BC083281.1 - 6 - 7 663 66
BC165788.1 - 6 - 7 685 66

View Full Product Details