Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rtn1b
Entrez Gene ID
327065
Gene Name
reticulon 1b
Gene Aliases
rtn1, rtn1l, wu:fa21b09, wu:fj35h01
Chromosome Location
Chr.20: 20637760 - 20695953 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029948.2 NP_001025119.1 2 - 3 426 74
NM_001328270.1 NP_001315199.1 2 - 3 595 74
NM_001328271.1 NP_001315200.1 4 - 5 2110 74
XM_005160490.4 XP_005160547.1 4 - 5 2213 74
GenBank mRNA AY555034.1 - 4 - 5 2110 74
AY555035.1 - 2 - 3 427 74
AY555036.1 - 2 - 3 415 74
BC052753.1 - 2 - 3 360 74
BC139604.1 - 2 - 3 360 74
BC153309.1 - 2 - 3 387 74

View Full Product Details