Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cacnb2b
Entrez Gene ID
558806
Gene Name
calcium channel, voltage-dependent, beta 2b
Gene Aliases
CACNB2.2, cacnb2a, im:7141271, si:dkey-32m20.2
Chromosome Location
Chr.2: 31462762 - 31539459 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081748.2 NP_001075217.2 - 848 70
NM_001166324.1 NP_001159796.1 - 606 70
GenBank mRNA BC160620.1 - 6 - 7 497 70
BC162962.1 - 6 - 7 464 70
BC162994.1 - 6 - 7 464 70
DQ372944.1 - 6 - 7 497 70
EU301442.1 - - 848 70
EU301443.1 - - 606 70

View Full Product Details