Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
taar1b
Entrez Gene ID
794715
Gene Name
trace amine associated receptor 1b
Gene Aliases
si:dkey-7c18.14
Chromosome Location
Chr.20: 46127591 - 46128590 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082904.1 NP_001076373.1 1 - 1 947 73
GenBank mRNA BC163111.1 - 1 - 1 1020 73
BC163112.1 - 1 - 1 1020 73

View Full Product Details