The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130935.3 | NP_571010.3 | 6 - 7 | 2368 | 60 |
GenBank mRNA | AF060117.1 | - | 6 - 7 | 2373 | 60 |
BC117641.1 | - | 6 - 7 | 2368 | 60 |