Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
wnt2bb
Entrez Gene ID
556087
Gene Name
wingless-type MMTV integration site family, member 2Bb
Gene Aliases
-
Chromosome Location
Chr.8: 718815 - 741357 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044344.1 NP_001037809.1 2 - 3 643 83
GenBank mRNA BC162931.1 - 2 - 3 669 83
BC162939.1 - 2 - 3 669 83
DQ231559.1 - 2 - 3 643 83

View Full Product Details