Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pcdh2ab12
Entrez Gene ID
493600
Gene Name
protocadherin 2 alpha b 12
Gene Aliases
zCNRv7ss-1, zcnrv12
Chromosome Location
Chr.14: 2123123 - 2187174 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009591.2 NP_001009591.2 1 - 1 1380 148
GenBank mRNA AB075940.1 - 1 - 1 1352 148

View Full Product Details