Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fundc1
Entrez Gene ID
436984
Gene Name
FUN14 domain containing 1
Gene Aliases
zgc:92600
Chromosome Location
Chr.9: 33783546 - 33785187 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002711.2 NP_001002711.2 2 - 3 217 107
GenBank mRNA BC076092.1 - 2 - 3 218 107
EH445440.1 - 2 - 3 226 107
EH467793.1 - 3 - 4 732 107

View Full Product Details