Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
angptl6
Entrez Gene ID
561927
Gene Name
angiopoietin-like 6
Gene Aliases
cb242, sb:cb242
Chromosome Location
Chr.3: 54524226 - 54550728 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014821.1 NP_001014821.1 4 - 5 1019 109
XM_009299794.3 XP_009298069.1 5 - 6 1344 109
GenBank mRNA AB199649.1 - 4 - 5 1019 109

View Full Product Details