Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ctsk
Entrez Gene ID
550475
Gene Name
cathepsin K
Gene Aliases
wu:fa95f03, wu:fb08b05, zgc:110367
Chromosome Location
Chr.16: 29492994 - 29502549 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017778.1 NP_001017778.1 3 - 4 335 93
GenBank mRNA BC092901.1 - 3 - 4 335 93
BC164579.1 - 2 - 3 291 93
HM239643.1 - 2 - 3 179 93
HM239644.1 - 2 - 3 179 93

View Full Product Details