Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tma16
Entrez Gene ID
553647
Gene Name
translation machinery associated 16 homolog
Gene Aliases
zgc:110664
Chromosome Location
Chr.1: 296568 - 301236 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177745.1 NP_001171216.1 5 - 6 490 95
GenBank mRNA BC095357.1 - 5 - 6 489 95
BC152117.1 - 5 - 6 449 95

View Full Product Details