Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
slc2a10
Entrez Gene ID
560546
Gene Name
solute carrier family 2 member 10
Gene Aliases
GLUT-10, zgc:171488
Chromosome Location
Chr.11: 2448789 - 2456840 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111163.1 NP_001104633.1 3 - 4 1389 129
XM_005172779.4 XP_005172836.1 4 - 5 1804 129
GenBank mRNA BC153938.1 - 3 - 4 1389 129
HM560591.1 - 4 - 5 1620 129
HM560592.1 - 3 - 4 1932 129

View Full Product Details