Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
march8
Entrez Gene ID
567818
Gene Name
membrane-associated ring finger (C3HC4) 8
Gene Aliases
zgc:171495
Chromosome Location
Chr.13: 17752591 - 18011412 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161435.2 NP_001154907.1 4 - 5 389 80
XM_009307238.3 XP_009305513.1 4 - 5 485 80
XM_009307239.3 XP_009305514.1 3 - 4 532 80
GenBank mRNA BC153940.1 - 3 - 4 344 80

View Full Product Details