Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
march8
Entrez Gene ID
567818
Gene Name
membrane-associated ring finger (C3HC4) 8
Gene Aliases
zgc:171495
Chromosome Location
Chr.13: 17752591 - 18011412 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC153940.1 - 5 - 6 661 101

View Full Product Details