Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ccdc85a
Entrez Gene ID
100141348
Gene Name
coiled-coil domain containing 85A
Gene Aliases
zgc:175167
Chromosome Location
Chr.13: 27000381 - 27038415 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_135163.1 2 - 3 2113 68
GenBank mRNA BC155335.1 - 2 - 3 2085 68

View Full Product Details