Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tmx1
Entrez Gene ID
323607
Gene Name
thioredoxin-related transmembrane protein 1
Gene Aliases
txndc, txndc1, wu:fc04g05
Chromosome Location
Chr.13: 36923099 - 36933317 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001115133.1 NP_001108605.1 2 - 3 318 77
GenBank mRNA BC056715.1 - 2 - 3 289 77
DR722002.1 - 2 - 3 337 77

View Full Product Details