Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cct6a
Entrez Gene ID
116994
Gene Name
chaperonin containing TCP1, subunit 6A (zeta 1)
Gene Aliases
fa95c01, wu:fa95c01
Chromosome Location
Chr.21: 41820359 - 41839678 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201290.1 NP_958447.1 6 - 7 772 68
XM_005161447.2 XP_005161504.1 7 - 8 1126 68
GenBank mRNA BC044393.1 - 6 - 7 772 68
BC071416.1 - 6 - 7 809 68
BC155176.1 - 7 - 8 1133 68

View Full Product Details