Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mtpn
Entrez Gene ID
406240
Gene Name
myotrophin
Gene Aliases
wu:fa10f07, zgc:64078
Chromosome Location
Chr.4: 1054708 - 1076053 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212972.2 NP_998137.1 3 - 4 313 70
XM_021474978.1 XP_021330653.1 3 - 4 797 70
GenBank mRNA BC054609.1 - 3 - 4 256 70
BC154009.1 - 3 - 4 280 70
BC155200.1 - 3 - 4 213 70
EH440876.1 - 3 - 4 313 70

View Full Product Details