Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
npsn
Entrez Gene ID
404039
Gene Name
nephrosin
Gene Aliases
npsnl, zgc:110470, zgc:152831
Chromosome Location
Chr.7: 38653481 - 38658412 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077779.2 NP_001071247.1 9 - 9 1046 107
GenBank mRNA BC095288.1 - 9 - 9 1044 107
BC124146.1 - 9 - 9 1044 107
EH434994.1 - 5 - 5 809 107

View Full Product Details