Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RSPH9
Entrez Gene ID
421446
Gene Name
radial spoke head component 9
Gene Aliases
C6orf206
Chromosome Location
Chr.3: 30921534 - 30942828 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277604.1 NP_001264533.1 3 - 4 541 143
NR_102410.1 3 - 4 477 143
XM_015283712.2 XP_015139198.1 3 - 4 517 143
GenBank mRNA BU492830.1 - 3 - 4 423 143
BX950488.1 - 3 - 4 541 143
CN227248.1 - 2 - 3 170 143

View Full Product Details