Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARL5B
Entrez Gene ID
420513
Gene Name
ADP ribosylation factor like GTPase 5B
Gene Aliases
ARL8
Chromosome Location
Chr.2: 19078341 - 19094395 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143969.1 NP_001137441.1 5 - 6 564 67
GenBank mRNA CR385805.1 - 5 - 6 564 67

View Full Product Details