Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
HACD1
Entrez Gene ID
420518
Gene Name
3-hydroxyacyl-CoA dehydratase 1
Gene Aliases
PTPLA
Chromosome Location
Chr.2: 19682110 - 19699098 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015281999.2 XP_015137485.1 6 - 7 802 145
XM_015282000.2 XP_015137486.1 6 - 7 808 145
GenBank mRNA CR386138.1 - 6 - 7 753 145
CR386940.1 - 6 - 7 760 145

View Full Product Details