Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PFDN4
Entrez Gene ID
419337
Gene Name
prefoldin subunit 4
Gene Aliases
-
Chromosome Location
Chr.20: 12335201 - 12337124 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278015.1 NP_001264944.1 3 - 4 324 81
GenBank mRNA BU114881.1 - 1 - 2 127 81
BX930309.1 - 3 - 4 350 81
BX935795.1 - 3 - 4 270 81
BX950659.1 - 3 - 4 274 81
CN226165.1 - 3 - 4 324 81

View Full Product Details