Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC420480
Entrez Gene ID
420480
Gene Name
Sjogren's syndrome nuclear autoantigen 1-like
Gene Aliases
SSNA1L
Chromosome Location
Chr.2: 15397367 - 15399485 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA CR352586.1 - 1 - 2 206 84

View Full Product Details