Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- AAED1
- Entrez Gene ID
-
427472
- Gene Name
- AhpC/TSA antioxidant enzyme domain containing 1
- Gene Aliases
-
C9ORF21, CZH9orf21
- Chromosome Location
-
Chr.Z: 42274779 - 42281198 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BX932659.1 |
- |
2 - 3 |
274 |
135 |
View Full Product Details