Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HNRNPLL
Entrez Gene ID
416702
Gene Name
heterogeneous nuclear ribonucleoprotein L like
Gene Aliases
HNRPLL
Chromosome Location
Chr.3: 16581734 - 16608471 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001305651.2 NP_001292580.2 3 - 4 673 84
GenBank mRNA BX930400.2 - 3 - 4 409 84
BX933171.1 - 3 - 4 673 84
CR388661.1 - 3 - 4 637 84

View Full Product Details