Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNTB1
Entrez Gene ID
404774
Gene Name
syntrophin beta 1
Gene Aliases
beta-1-syntrophin
Chromosome Location
Chr.2: 136964312 - 137072270 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005798.1 NP_001005798.1 1 - 2 576 64
XM_015283067.2 XP_015138553.2 1 - 2 645 64
GenBank mRNA AB105810.1 - 1 - 2 275 64
AJ719936.1 - 1 - 2 576 64
CR388496.1 - 1 - 2 196 64

View Full Product Details