Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MED20
Entrez Gene ID
419926
Gene Name
mediator complex subunit 20
Gene Aliases
TRFP
Chromosome Location
Chr.26: 5021518 - 5026435 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006327.2 NP_001006327.1 - 186 55
GenBank mRNA AJ719945.1 - - 188 55
CR354350.1 - - 302 55

View Full Product Details