Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTPN
Entrez Gene ID
395702
Gene Name
myotrophin
Gene Aliases
-
Chromosome Location
Chr.1: 58205713 - 58243481 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204886.2 NP_990217.1 1 - 2 395 68
GenBank mRNA AJ445794.1 - 1 - 2 396 68
AJ720711.1 - 1 - 2 262 68
AJ721118.1 - 1 - 2 291 68
D26326.1 - 1 - 2 205 68

View Full Product Details