Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JCHAIN
Entrez Gene ID
374117
Gene Name
joining chain of multimeric IgA and IgM
Gene Aliases
IGJ
Chromosome Location
Chr.4: 50012276 - 50018044 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204263.1 NP_989594.1 2 - 3 342 118
GenBank mRNA AB025103.1 - 2 - 3 342 118

View Full Product Details