Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UMOD
Entrez Gene ID
404754
Gene Name
uromodulin
Gene Aliases
ZPD
Chromosome Location
Chr.11: 523817 - 527808 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213576.2 NP_998741.2 8 - 9 1182 74
XM_015292282.2 XP_015147768.1 7 - 8 1170 74
XM_015292283.2 XP_015147769.1 7 - 8 1153 74
GenBank mRNA AB114441.2 - 8 - 9 1182 74

View Full Product Details