Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDUFA10
Entrez Gene ID
424032
Gene Name
NADH:ubiquinone oxidoreductase subunit A10
Gene Aliases
-
Chromosome Location
Chr.7: 6553073 - 6593673 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031247.1 NP_001026418.1 8 - 9 939 82
XM_015289173.2 XP_015144659.1 8 - 9 971 82
GenBank mRNA AJ720729.1 - 8 - 9 939 82

View Full Product Details