Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NFE2L1
Entrez Gene ID
417987
Gene Name
nuclear factor, erythroid 2 like 1
Gene Aliases
-
Chromosome Location
Chr.27: 6479451 - 6488365 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030756.1 NP_001025927.1 5 - 6 1021 63
GenBank mRNA AJ719867.1 - 5 - 6 1021 63
CR354056.1 - 3 - 4 755 63

View Full Product Details