Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSHR
Entrez Gene ID
428900
Gene Name
thyroid stimulating hormone receptor
Gene Aliases
-
Chromosome Location
Chr.5: 40973010 - 41024583 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033850.1 NP_001029022.1 6 - 7 570 106
NM_001193588.1 NP_001180517.1 8 - 9 717 106
NM_001193589.1 NP_001180518.1 7 - 8 642 106
XM_025150837.1 XP_025006605.1 8 - 9 900 106
GenBank mRNA AB234613.1 - 8 - 9 717 106
AB234614.1 - 7 - 8 642 106
AB234615.1 - 6 - 7 570 106
AB376101.1 - 1 - 2 75 106

View Full Product Details