Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BG-like antigen 1
Entrez Gene ID
426940
Gene Name
BG1
Gene Aliases
B-G, BG-IV
Chromosome Location
Chr.16: 2549788 - 2554982 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099355.1 NP_001092825.1 3 - 4 546 77
XM_015294968.2 XP_015150454.1 3 - 4 1051 77
XM_015294971.2 XP_015150457.1 3 - 4 1051 77
XM_015294972.2 XP_015150458.1 3 - 4 1052 77
XM_015294975.2 XP_015150461.1 3 - 4 1050 77
XM_015294976.2 XP_015150462.1 3 - 4 1052 77
XM_015294977.2 XP_015150463.1 3 - 4 1048 77
XM_015294978.2 XP_015150464.1 3 - 4 1050 77
XM_015294979.2 XP_015150465.1 3 - 4 1049 77
XM_015294980.2 XP_015150466.1 3 - 4 1050 77
XM_025155763.1 XP_025011531.1 3 - 4 1050 77
XM_025155765.1 XP_025011533.1 3 - 4 1047 77
GenBank mRNA KC955132.1 - 3 - 4 1036 77
KC955133.1 - 4 - 5 609 77
KC955135.1 - 4 - 5 908 77
KX686501.1 - 3 - 4 778 77
KX686503.1 - 3 - 4 781 77

View Full Product Details