Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BG-like antigen 1
Entrez Gene ID
426940
Gene Name
BG1
Gene Aliases
B-G, BG-IV
Chromosome Location
Chr.16: 2549788 - 2554982 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099355.1 NP_001092825.1 7 - 8 630 118
XM_015294968.2 XP_015150454.1 7 - 8 1135 118
XM_015294970.2 XP_015150456.1 7 - 8 1131 118
XM_015294971.2 XP_015150457.1 7 - 8 1135 118
XM_015294972.2 XP_015150458.1 7 - 8 1136 118
XM_015294973.2 XP_015150459.1 6 - 7 1114 97
XM_015294974.2 XP_015150460.1 6 - 7 1110 97
XM_015294977.2 XP_015150463.1 7 - 8 1132 118
XM_015294979.2 XP_015150465.1 7 - 8 1133 118
XM_015294981.2 XP_015150467.1 7 - 8 1129 118
GenBank mRNA JQ780447.1 - 7 - 8 627 118
JQ780448.1 - 7 - 8 627 118
KC955132.1 - 7 - 8 1120 118
KC955133.1 - 8 - 9 693 118
KC955134.1 - 8 - 9 690 118
KC955135.1 - 8 - 9 992 118
KC955136.1 - 7 - 8 1117 118
KX686501.1 - 7 - 8 862 118
KX686502.1 - 7 - 8 865 118
KX686503.1 - 7 - 8 865 118
KX686504.1 - 7 - 8 863 118
KX686505.1 - 7 - 8 860 118

View Full Product Details