Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMO2
Entrez Gene ID
374129
Gene Name
LIM domain only 2
Gene Aliases
rhombotin-2
Chromosome Location
Chr.5: 18658099 - 18665579 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204271.1 NP_989602.1 2 - 3 301 86
XM_015286755.2 XP_015142241.1 3 - 4 685 86
GenBank mRNA AF468789.1 - 2 - 3 301 86

View Full Product Details