Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MKRN2
Entrez Gene ID
374162
Gene Name
makorin ring finger protein 2
Gene Aliases
-
Chromosome Location
Chr.12: 5341076 - 5354439 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204288.1 NP_989619.1 5 - 6 871 71
XM_015292934.2 XP_015148420.1 5 - 6 854 71
XM_015292935.1 XP_015148421.1 5 - 6 854 71
XM_025154402.1 XP_025010170.1 5 - 6 854 71
XM_025154403.1 XP_025010171.1 5 - 6 907 71
GenBank mRNA AF439559.1 - 5 - 6 871 71

View Full Product Details