Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAV3
Entrez Gene ID
378796
Gene Name
caveolin 3
Gene Aliases
caveolin-3
Chromosome Location
Chr.12: 19676690 - 19680119 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204370.2 NP_989701.2 1 - 2 194 79
GenBank mRNA AY363370.1 - 1 - 2 119 79
BX931541.1 - 1 - 2 202 79
BX932348.2 - 1 - 2 190 79
BX934028.1 - 1 - 2 148 79

View Full Product Details