Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HMCES
Entrez Gene ID
416015
Gene Name
5-hydroxymethylcytosine binding, ES cell specific
Gene Aliases
C12H3ORF37, C3orf37, DC12, SRAPD1
Chromosome Location
Chr.12: 9520448 - 9523465 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006137.1 NP_001006137.1 4 - 5 660 57
XM_015293076.2 XP_015148562.1 3 - 4 551 57
GenBank mRNA AJ720353.1 - 4 - 5 660 57

View Full Product Details