Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BZW1
Entrez Gene ID
424073
Gene Name
basic leucine zipper and W2 domains 1
Gene Aliases
-
Chromosome Location
Chr.7: 11284764 - 11294541 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006516.1 NP_001006516.1 11 - 12 1345 104
XM_025152181.1 XP_025007949.1 9 - 10 104
GenBank mRNA AJ719647.1 - 11 - 12 1345 104

View Full Product Details